Safety gas burners, JUMBO, with stopcock
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7511 7512
BOCH7511EA 609 EUR
BOCH7511 BOCH7512
Safety gas burners, JUMBO, with stopcock
Verwarmers Gasbranders
Safety gas burner JUMBO with stopcock for natural gas/propane.
  • Thermo safety element
  • Bimetal-flame antena magnet breech and pilot flame
  • Air regulation
  • Natural gas and propane gas/butane gas
  • Max. 1300 °C