Buretklemmen
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5476 5477
BOCH5476EA 45.6 EUR
BOCH5476 BOCH5477
Buretklemmen
Klemmen Klemmen voor algemeen gebruik
Burette clamps made of zinc plated stainless steel.
  • Fingers are covered with silicone coating to protect clamped items
  • Single sided or double sided available