Allylbenzene ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
1-Phenyl-2-propene, 3-Phenylpropene
A12965.14
A12965.22
A12965.14EA
60.3
EUR
A12965.14
A12965.22
Allylbenzene ≥98%
Allylbenzene
Purity: 98%
CAS No.: 300-57-2
Molecular Formula: C9H10
Formula Weight: 118.18
Flash Point: 33°(91°F)
Refractive Index: 1.5100
Odour: Aromatic
Melting Point: ca -50°
Boiling Point: 158-160°
Beilstein Reference: 1098501
UN No.: 3295
MDL No.: MFCD00008651
Formule:
(C₆H₅)CH₂CHCH₂ Kookpunt: 158…160 °C Smeltpunt: ∼–50 °C Dichtheid: 0,892 Vlampunt: 33 °C (91 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00008651 CAS nummer: 300-57-2 EINECS: 206-095-7 UN: 3295 ADR: 3,III |