Retort clamps, with flexible shaft
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B08303033 5830 5850 5840 B08309658
451-9110EA 180 EUR
451-9110 241-3135 241-3137 241-3136 BOCHB08309658
Retort clamps, with flexible shaft
Klemmen Klemmen voor algemeen gebruik
Retort clamps with flexible shaft, either made of stainless steel or an aluminium alloy (Laboral).
  • Finger with silicone coating
  • Shaft nickel plated