Clamps, three pronged, with shaft
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5627 5626
BOCH5627EA 37.7 EUR
BOCH5627 BOCH5626
Clamps, three pronged, with shaft
Klemmen Klemmen voor algemeen gebruik
Chromium plated three-finger clamps with PVC coated fingers. Span width independently adjustable by two regulating screws.