Aansluitbare schroefdraadbuizen, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286063403 286253201 286256207 286258808 286251603
SCOT286063403EA 165 EUR
SCOT286063403 SCOT286253201 SCOT286256207 SCOT286258808 SCOT286251603
Aansluitbare schroefdraadbuizen, DURAN®
Adapters Adapters, Buizen met schroefdraad
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread GL. It is used by glassblowers in the construction of laboratory glassware.
Opgelet: These tubes are open at the bottom.