Cryovials, Cryovial®
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T309-1A T309-2 T309-2A T309-3A T309-4A T309-5A T310-10A T310-1A T310-2 T310-3A T310-4A T311-1 T311-2 T311-3 T311-4 T311-4A T311-5 T310-5A T310-2A T308-3A T301-1 T308-2A T310-10AES
479-4507EA 586 EUR
479-4507 479-0080 720-0997 479-0081 720-0998 720-0999 479-0082 720-1000 479-0083 525-0210 479-0084 720-1003 720-1004 720-1005 720-1006 479-0085 479-0096 720-1002 720-1001 SIMPT308-3A 479-1375 SIMPT308-2A SIMPT310-10AES
Cryovials, Cryovial®
flesjes
PP vial and closure. Designed for the storage of biological material, human or animal cells, at temperatures as low as −196 °C (but should be used only in the gas phase of liquid nitrogen).
  • Certified RNase-, DNase-, pyrogen- and DNA-free
  • With graduations and marking area
Round bottom vials can be centrifuged up to 17000×g
Round bottom vials can be centrifuged up to 17 000 ×g