Bolvormige slijpingen, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286320602 286321607 286322406 286323205 286324004 286324801 286326005 286326802 286327601 286327807 286328606 286329602
SCOT286320602EA 646 EUR
SCOT286320602 SCOT286321607 SCOT286322406 SCOT286323205 SCOT286324004 SCOT286324801 SCOT286326005 SCOT286326802 SCOT286327601 SCOT286327807 SCOT286328606 SCOT286329602
Bolvormige slijpingen, DURAN®
Adapters Adapters met kogelgewrichten
DURAN®, borosilicate glass 3.3, clear.
  • Inches size
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Through use of flexible spherical joint connections, apparatus can be assembled free from stresses.
*non-DIN size
Certificaten: DIN 12244, DIN 12264