Bolvormige slijpingen, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286321401 286323008 286324604 286325806 286329405 286327404
SCOT286321401EA 192 EUR
SCOT286321401 SCOT286323008 SCOT286324604 SCOT286325806 SCOT286329405 SCOT286327404
Bolvormige slijpingen, DURAN®
Adapters Adapters met kogelgewrichten
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ungrounded, for glass manipulators.
Order Now
Specificaties
Environmentally Preferable