Bolvormige slijpingen, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286323608 286325206 286326408 286328006 286329808 286322003
SCOT286323608EA 196 EUR
SCOT286323608 SCOT286325206 SCOT286326408 SCOT286328006 SCOT286329808 SCOT286322003
Bolvormige slijpingen, DURAN®
Adapters Adapters met kogelgewrichten
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ungrounded, for glass manipulators.