Aansluitbare schroefdraadbuizen, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286252205 286254009 286256807 286259607 286263005
SCOT286252205EA 2.4 EUR
SCOT286252205 SCOT286254009 SCOT286256807 SCOT286259607 SCOT286263005
Aansluitbare schroefdraadbuizen, DURAN®
Adapters Adapters, Buizen met schroefdraad
Made of DURAN®, borosilicate glass 3.3, clear.
  • heavy wall
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread GL. It is used by glassblowers in the construction of laboratory glassware.
Supplementary to DIN 12216