Stirrers, propeller, with four paddles
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9054 9053 9055 9056 9057 9058
BOCH9054EA 37.2 EUR
BOCH9054 441-1900 BOCH9055 BOCH9056 BOCH9057 BOCH9058
Stirrers, propeller, with four paddles
Roerders Roergereedschap
Propeller stirrer made of stainless steel with four vertical paddles.
  • Mixing of media with low and middle viscosity
  • Radial flow