Stirrers, propeller, with four paddles
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9003 9009 9010 9011 9004 9005 9012 9013 9014
BOCH9003EA 31 EUR
BOCH9003 BOCH9009 BOCH9010 BOCH9011 BOCH9004 BOCH9005 BOCH9012 BOCH9013 BOCH9014
Stirrers, propeller, with four paddles
Roerders Roergereedschap
Propeller stirrer made of stainless steel with four paddles.
  • Mixing of media with low and middle viscosity
  • Ideal for homogenisation
  • Axial flow