Stirrers, propeller, with three paddles
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9109 9112 9114 9103 9110 9111 9113 9104 9105 9115 9116
BOCH9109EA 31 EUR
BOCH9109 BOCH9112 441-1904 BOCH9103 BOCH9110 BOCH9111 BOCH9113 BOCH9104 BOCH9105 BOCH9115 BOCH9116
Stirrers, propeller, with three paddles
Roerders Roergereedschap
Propeller stirrer made of stainless steel with three paddles.
  • Mixing of media with low and middle viscosity
  • Ideal for homogenisation
  • Axial flow