Overhead stirrer tools, anchor stirrers
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9611 9610 9612
BOCH9611EA 112 EUR
BOCH9611 BOCH9610 BOCH9612
Overhead stirrer tools, anchor stirrers
Roerders Roergereedschap
Anchor stirrers made of stainless steel.
  • Ideal for stirring liquids at low speeds
  • Suitable for stirring high viscous medium
  • Creates radial flow