Isododecane (mixture of isomers) ≥80%, tech.
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Isododecane, mixture of isomer acts as a reaction medium used for polymerisation reactions. It is also useful as an emollient and solvent in skin care products due to its high spreadability, low viscosity and density. It is commonly used in anti-aging serums and also useful in many different cosmetic items like eyeliner, hair care, hair sprays, perfume, conditioners and lotions.
Purity: 80%
CAS No.: 31807-55-3
Molecular Formula: C12H26
Formula Weight: 170.34
Flash Point: 42°(108°F)
Physical Form: Liquid
Density: 0.74
Refractive Index: 1.4210
Appearance: Colourless
Boiling Point: 170-195°
UN No.: 3295
MDL No.: MFCD00036106
Opgelet: Lab use only.
Formule:
C₁₂H₂₆ MW: 170.34 g/mol Kookpunt: 170…195 °C Dichtheid: 0.74 Vlampunt: 42 °C (108 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00036106 CAS nummer: 31807-55-3 EINECS: 250-816-8 UN: 2286 ADR: 3 III |