(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: (S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, (2S)-(-)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid
B23117.88 B23117.ME
B23117.88EA 331 EUR
B23117.88 B23117.ME
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥99%
(S)-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid
Purity: 99% CAS No.: 17257-71-5 Molecular Formula: C10H9F3O3 Formula Weight: 234.18 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 43-45° Boiling Point: 105-107°/1mm Beilstein Reference: 3591561 Merck Reference: 14,6280 Optical Rotation: -72° (c=2 in methanol) MDL No.: MFCD00064200
Formule: C₁₀H₉F₃O₃
MW: 234.17 g/mol
Kookpunt: 105…107 °C (1 mmHg)
Smeltpunt: 43…45 °C
Dichtheid: 1.344
Vlampunt: >110 °C (230 °F)
MDL Nummer: MFCD00064200
CAS nummer: 17257-71-5
EINECS: 241-292-1