Dihydromyrcenol, liquid

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

0215090680 0215090625
ICNA0215090680EA 188 EUR
ICNA0215090680 ICNA0215090625
Dihydromyrcenol, liquid
Dihydromyrcenol
Formule: C₁₀H₂₀O
CAS nummer: 18479-58-8
EINECS: 242-362-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR