Supelco® Organic Reference Standard, Myrcene
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
64643-100MG-F 64643-500MG-F
64643-100MG-FEA 46.7 EUR
64643-100MG-F 64643-500MG-F
Supelco® Organic Reference Standard, Myrcene
Standaarden Organische standaarden
  • GHS signal: GHS02, GHS08, GHS07
Opgelet: P210 - P273 - P301 + P310 - P303 + P361 + P353 - P305 + P351 + P338 - P331
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06488246b5b4e63a4bbd0377844c0ca70abdb97d&vid=06488246b5b4e63a4bbd0377844c0ca70abdb97d&prid=web&clid=SAPDAM
Order Now
Specificaties
Synonym(s) β-Myrcene; 7-Methyl-3-methylene-1,6-octadiene
M-Clarity quality level MQ100
Environmentally Preferable
Dichtheid 0,791 g/ml at 25 °C (lit.)
CAS No. 123-35-3
MDL No. MFCD00008908
Formule H₂C=CHC(=CH₂)CH₂CH₂CH=C(CH₃)₂
Moleculair gewicht 136,23
Kookpunt 167 °C (lit.)
Beschrijving van de gevarenklasse H226 - H304 - H315 - H319 - H410
Vlampunt 44 °C - closed cup
Shipping conditions STD
Storage conditions –20 °C