Overhead stirrer tools, paddle stirrers
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9503 9507 9504 9505 9506 9508
441-1906EA 39.2 EUR
441-1906 BOCH9507 BOCH9504 BOCH9505 BOCH9506 441-1907
Overhead stirrer tools, paddle stirrers
Roerders Roergereedschap
Paddle stirrers made of stainless steel, with six holes.
  • Ideal for stirring liquids at middle speed range
  • Suitable for stirring low to medium viscous media
  • Creates radial flow