Overhead stirrer tools, centrifugal stirrers
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9210 9211 9212 9213 9214 9209 9215 9216
BOCH9210EA 31 EUR
BOCH9210 BOCH9211 BOCH9212 BOCH9213 BOCH9214 441-1905 BOCH9215 BOCH9216
Overhead stirrer tools, centrifugal stirrers
Roerders Roergereedschap
Centrifugal stirrers made of stainless steel, with two flexible blades.
  • Ideal for stirring liquids at middle speed range
  • Suitable for stirring low to medium viscous media
  • Creates radial flow
  • Suitable for jars with tight neck
Order Now
Specificaties
Environmentally Preferable