Overhead stirrer tools, viscosity stirrers, Visco-Jet®
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9020 9021
BOCH9020EA 1300 EUR
BOCH9020 BOCH9021
Overhead stirrer tools, viscosity stirrers, Visco-Jet®
Roerders Roergereedschap
Viscosity stirrers made of stainless steel.
  • Ideal for stirring liquids at low speed
  • Suitable for stirring low and highly viscous media
  • Creates dynamic radial flow
Complete degasification of the media, foam generation and air inlet will be avoided.