PCR-platen, 96-wells, verhoogde rand, Amplate™
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
T323-103N T323-100N T323-101N
SIMPT323-103NEA 469 EUR
SIMPT323-103N SIMPT323-100N SIMPT323-101N
PCR-platen, 96-wells, verhoogde rand, Amplate™
Microplaten PCR-platen
Amplate™ raised rim, thin walled PCR plates offer the rigidity needed for automation and are perfectly suited for high performance thermal cycling. Each well makes intimate contact with the heating block while quick and consistent heat transfer is ensured by a uniform wall thickness.
  • PP
  • Using polypropylene with low adhesion surface properties ensures samples are easily recovered
  • An alphanumeric grid helps sample identification
All sealing methods can be used for oil-free operation: Domed and flat cap strips; SecureSeal™ Thermal Sealing Film; SecureSeal™ Aluminium Sealing Foil and Amplate™ Mat. To facilitate orientation, one corner of the plate is cut away.
Leveringsinformatie: Packed in tamperproof resealable bags of ten plates.