HPLC-kolommen, Accucore™ aQ
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17326-012105 17326-013005 17326-014005 17326-052130 17326-053030 17326-102130 17326-103030 17326-104630 17326-152130 17326-154630
554-1948EA 515 EUR
554-1948 554-1949 554-1950 554-1897 554-1898 554-1900 554-1901 554-1902 554-1903 554-1905
HPLC-kolommen, Accucore™ aQ
Kolommen voor chromatografie
These columns are compatible with 100% aqueous mobile phases and offer special selectivity for polar analytes. Based on Core Enhanced Technology, they provide fast, high resolution separations without the elevated back pressures required by sub 2 µm particles.
  • Ideal for highly aqueous mobile phases
  • Retention and resolution of polar analytes
  • Polar endcapped C18 stationary phase for alternative selectivity