HPLC-kolommen, Accucore™ Phenyl-Hexyl
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17926-012105 17926-013005 17926-014005 17926-052130 17926-053030 17926-054630 17926-102130 17926-103030 17926-104630 17926-152130 17926-153030 17926-154630
554-1951EA 526 EUR
554-1951 554-1952 554-1953 554-1909 554-1910 554-1911 554-1912 554-1913 554-1914 554-1915 554-1916 554-1917
HPLC-kolommen, Accucore™ Phenyl-Hexyl
Kolommen voor chromatografie
These columns offer unique selectivity for aromatic and moderately polar analytes. Suitable for complex samples where some peaks are well resolved on a phenyl phase whilst others require a conventional alkyl phase. Based on Core Enhanced Technology, they provide fast, high resolution separations without the elevated back pressures required by sub-2 µm particles.
  • Lower back pressures
  • Mixed-mode selectivity for aromatic and moderately polar analytes
  • Enhanced pi-pi interactions with aromatics
  • Moderate hydrophobicity