HPLC-kolommen, Accucore™ HILIC
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17526-012105 17526-013005 17526-014005 17526-052130 17526-102130 17526-152130 17526-153030 17526-154630
554-1957EA 602 EUR
554-1957 554-1958 554-1959 554-1933 554-1936 554-1939 554-1940 554-1941
HPLC-kolommen, Accucore™ HILIC
Kolommen voor chromatografie
These columns offer enhanced retention of polar and hydrophilic analytes. Based on Core Enhanced Technology, they provide fast, high resolution separations without the elevated back pressures required by sub-2 µm particles.
  • Lower back pressure
  • Alternative selectivity to C18 without ion-pair or derivatisation
  • Improved sensitivity for MS detection
Rugged 2.6 µm solid core particles enable fast, high-resolution separations at low backpressures. The HILIC stationary phase provides alternative selectivity to C18 without ion-pairing or derivatization. TheMore Product Information