4-Bromobenzyl chloride 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
346750050 346750250
ACRO346750050EA 49.5 EUR
ACRO346750050 ACRO346750250
4-Bromobenzyl chloride 98%
4-Bromobenzyl chloride
Formule: C₇H₆BrCl
Kookpunt: 136…139 °C (27 mmHg)
Smeltpunt: 36…40 °C
MDL Nummer: MFCD00040867
CAS nummer: 589-17-3
EINECS: 209-638-6
UN: 3261
ADR: 8,II
Merck Index: 13,01394
Order Now
Specification Test Results
Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 36 to 40 °C
GC ≥97.5 %