Glass cutters
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
12250 12251
BOCH12250EA 104 EUR
BOCH12250 BOCH12251
Glass cutters
Snijders
Plastic handle with hardened metal blade.
  • Clean and precise cutting
  • Replaceable hardened metal blades
  • Easy to change blade
Length 180 mm, blade dimensions 40×18 mm.