Emmers, met maatverdeling, met tuit, roestvrij staal 18/10
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8390 8391 8394
BOCH8390EA 1040 EUR
BOCH8390 BOCH8391 BOCH8394
Emmers, met maatverdeling, met tuit, roestvrij staal 18/10
Emmers
Stainless steel 18/10.
  • Very good chemical resistance, very good thermal resistance
  • High quality, very stable
  • Matching lids available optionally
For use in laboratories and industry. Graduated with handle. Seamless handle tabs and smooth surfaces for fast, easy cleaning.