α-Bromo-4-nitrotoluene

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

SB00745.1 SB00745.10 SB00745.50
MAYBSB00745.1EA 0 EUR
MAYBSB00745.1 MAYBSB00745.10 MAYBSB00745.50
α-Bromo-4-nitrotoluene
α-Bromo-4-nitrotoluene
Formule: C₇H₆BrNO₂
Kookpunt: 304 °C (1013 hPa)
Smeltpunt: 94…96 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00007373
CAS nummer: 100-11-8
EINECS: 202-820-6
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR