4-Tolualdehyde ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
p-Tolualdehyde, p-Tolylaldehyde, p-Methylbenzaldehyd, 4-Methylbenzaldehyd, 4-Methylbenzaldehyde, 4-Tolylaldehyde
A11384.22
A11384.06
A11384.0E
A11384.36
A11384.22EA
29.8
EUR
A11384.22
A11384.06
A11384.0E
A11384.36
4-Tolualdehyde ≥98%
4-Tolualdehyde
Purity: 98%
CAS No.: 104-87-0
Molecular Formula: C8H8O
Formula Weight: 120.15
Flash Point: 80°(176°F)
Refractive Index: 1.5460
Melting Point: -6°
Boiling Point: 204°
Beilstein Reference: 385772
MDL No.: MFCD00006954
Formule:
C₈H₈O MW: 120.15 g/mol Kookpunt: 204 °C Smeltpunt: –6 °C Dichtheid: 1.018 Vlampunt: 80 °C (176 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006954 CAS nummer: 104-87-0 EINECS: 203-246-9 |