Flow meters
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5700-0141 5700-0121 5700-0111
BURK5700-0141EA 1170 EUR
BURK5700-0141 BURK5700-0121 BURK5700-0111
Flow meters
Debietmeters
These flow meters are compact and fully electronic for electric drum and container pumps, also used for low viscosity media.
  • Measurement by the turbine wheel principle
  • Can be calibrated depending on the medium
  • Different versions for many different applications
  • Automatic switch on when flow is recognized
  • Four resettable part ranges and one fixed total range