4-Hydroxybenzylacetone

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Raspberry ketone, 1-(4-Hydroxyphenyl)-3-butanone, 4-(4-Hydroxyphenyl)-2-butanone

SB01721.1 SB01721.50
MAYBSB01721.1EA 0 EUR
MAYBSB01721.1 MAYBSB01721.50
4-Hydroxybenzylacetone
4-Hydroxybenzylacetone
Formule: C₁₀H₁₂O₂
Kookpunt: 292 °C (1013 hPa)
Smeltpunt: 83…84 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002394
CAS nummer: 5471-51-2
EINECS: 226-806-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR