Draadmanden voor transport
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17300 17301 17302 17305 17306 17307 17310 17311 17312
BOCH17300EA 502 EUR
BOCH17300 BOCH17301 BOCH17302 BOCH17305 BOCH17306 BOCH17307 BOCH17310 BOCH17311 BOCH17312
Draadmanden voor transport
Manden Draadmanden
Stainless steel 18/10.
  • Three different mesh dimensions
  • Stackable
  • Excellent thermal and chemical resistance
These rectangular baskets are designed for transporting, storing, cleaning and sterilising laboratory utensils.