6-Methylanthranil acid
Leverancier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2-Amino-6-methylbenzoic acid
SB00723.1
SB00723.10
SB00723.50
MAYBSB00723.1EA
0
EUR
MAYBSB00723.1
MAYBSB00723.10
MAYBSB00723.50
6-Methylanthranil acid
6-Methylanthranil acid
Formule:
C₈H₉NO₂ MW: 151.16 g/mol Kookpunt: 315 Smeltpunt: 125…127 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00007809 CAS nummer: 4389-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...