α,α,α,4-Tetrafluorotoluene
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SB00687.1 SB00687.10 SB00687.50
MAYBSB00687.1EA 0 EUR
MAYBSB00687.1 MAYBSB00687.10 MAYBSB00687.50
α,α,α,4-Tetrafluorotoluene
α,α,α,4-Tetrafluorotoluene
Formule: C₇H₄F₄
Kookpunt: 102 °C (1013 hPa)
Dichtheid: 1,293 g/cm³ (25 °C)
MDL Nummer: MFCD00000399
CAS nummer: 402-44-8
EINECS: 206-944-1
UN: 1993
ADR: 3,II