Adapter, conisch geslepen verbinding op schroefdraad, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
248417105 248409208 248416109 248418101 248419106 248427209 248428205 248429201 248448104 248449109 248406202 248407207 248408203
SCOT248417105EA 170 EUR
SCOT248417105 SCOT248409208 SCOT248416109 SCOT248418101 SCOT248419106 SCOT248427209 SCOT248428205 SCOT248429201 SCOT248448104 SCOT248449109 SCOT248406202 SCOT248407207 SCOT248408203
Adapter, conisch geslepen verbinding op schroefdraad, DURAN®
Adapters Adapters met NS-verbindingen
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Threaded adapter with DIN thread and standard ground cone.
DIN 12257
* non-DIN size