Filter tips, aeroject® ultra
Leverancier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1770010 1770020 1770100 1770200
RATI1770010EA 128 EUR
RATI1770010 RATI1770020 RATI1770100 RATI1770200
Filter tips, aeroject® ultra
Pipettips
These sterile pipette tips with filters effectively prevent the pipettor shaft from contamination with aerosols and liquids.
  • Free of RNase-, DNase-, DNA-, pyrogens, ATP
  • Free of human DNA and PCR inhibitors
  • Tips and aerosol filters made of ultra-hydrophobic PE-HD
  • Minimised retention due to special surface treatment
  • Perfect and tight fit on all major pipettor brands
  • Tips with extended length: Ideal for operation with narrow vessels and reduced risk of cross-contamination
Verpakking: Packed in plastic racks with hinged lids.