4-Fluoro-2-methylphenol ≥98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
A16032.06
A16032.14
A16032.22
A16032.06EA
46.8
EUR
A16032.06
A16032.14
A16032.22
4-Fluoro-2-methylphenol ≥98%
4-Fluoro-2-methylphenol
Purity: 98%
CAS No.: 452-72-2
Molecular Formula: C7H7FO
Formula Weight: 126.13
Flash Point: 85°(185°F)
Refractive Index: 1.5165
Odour: Phenol-like
Melting Point: 32-36°
Boiling Point: 87°/14mm
Beilstein Reference: 2041490
UN No.: 3261
MDL No.: MFCD00075088
Formule:
C₇H₇FO Kookpunt: 87 °C (14 mmHg) Smeltpunt: 32…36 °C Vlampunt: 85 °C (185 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00075088 CAS nummer: 452-72-2 UN: 3261 ADR: 8,III |