Disposable cuvettes
Leverancier: RATIOLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2711110 2712120
RATI2711110EA 115 EUR
RATI2711110 RATI2712120
Disposable cuvettes
Cuvetten Cuvetten voor spectrofotometers
These cuvettes with reduced wall thickness result in increased heat transfer.
  • Very low variation of extinction values
Verpakking: In PS boxes with lid, 100 cuvettes per box.