Afdichtingen van siliconenrubber DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286200405 286200602 286200808 286201007 286201204 286201401 286201607 286204604 286217602 286217808 286218007 286218204 286218401 286218607 286204801
SCOT286200405EA 16.3 EUR
SCOT286200405 SCOT286200602 SCOT286200808 SCOT286201007 SCOT286201204 SCOT286201401 SCOT286201607 SCOT286204604 SCOT286217602 SCOT286217808 SCOT286218007 SCOT286218204 SCOT286218401 SCOT286218607 SCOT286204801
Afdichtingen van siliconenrubber DURAN®
Afdichtingen Afdichtringen
Silicone, translucent.
  • Without washer
  • Very good chemical resistance
  • Very good temperature resistance