Statiefkoppelingen, driehoekig
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5655 5656
BOCH5655EA 17.5 EUR
BOCH5655 BOCH5656
Statiefkoppelingen, driehoekig
Klemmen Statiefkoppelingen
Three angled bossheads made of duraluminium (aluminium alloy).
  • Angle: 90 + 180 + 270°
  • Suitable for multilevel and three dimension constructions