Double bossheads, cross type
Leverancier: BOCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5306 5366
BOCH5306EA 90.7 EUR
BOCH5306 BOCH5366
Double bossheads, cross type
Klemmen Statiefkoppelingen
Cross type double bossheads, either made of stainless steel or malleable cast iron, chromium plated.
  • For rods up to 16,5 mm
  • Angle: 90 + 180°
  • With safety screw type 'heart' (M6)