Stirring paddles, PP
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8203-2100 8203-3150
BURK8203-2100EA 63.8 EUR
BURK8203-2100 BURK8203-3150
Stirring paddles, PP
Roerders Roergereedschap
PP stirring paddles ideal for gentle stirring in plastic and glass containers.
  • Shatterproof
  • Warp resistant