Tubing clamps, Squeeze-Fix
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8618-0002 8618-0003 8618-0001
BURK8618-0002EA 1.7 EUR
BURK8618-0002 BURK8618-0003 BURK8618-0001
Tubing clamps, Squeeze-Fix
Klemmen Slangklemmen
Permanently elastic hose clamps made of white POM.
  • Hose can be squeezed in phases or completely compressed, as required
  • Can be operated with one hand
  • Sterilisable, do not rust