Stopcocks, straight or multi-way bore
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8610-0012 8610-0015 8610-0003 8610-0006 8610-0001 8610-0010
BURK8610-0012EA 43.8 EUR
BURK8610-0012 BURK8610-0015 BURK8610-0003 BURK8610-0006 BURK8610-0001 BURK8610-0010
Stopcocks, straight or multi-way bore
Afsluitkranen
PTFE.
  • Resistant to compression max. 1 bar
  • Vacuum suitable
  • Autoclavable