Transport trolleys
Leverancier: HUPFER METALLWERKE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0114192 0114212 0112662 0112692
139-9902EA 1480 EUR
139-9902 139-9903 139-9904 139-9906
Transport trolleys
Transportwagens
These trolleys are made from high quality stainless steel.
Platform trolley: Flat metal platform, folded down by 80 mm all around with two plastic bumpers at the front. It has four swivel castors, two of them with total brake, Ø 125 mm
Single-ended trolley: Platform manufactured from round tube, Ø 25 mm, with welded-on stainless steel plate. It has two swivel castors with total brake, two fixed castors, Ø 200 mm