Onderzoekshandschoenen, latex, sterile, Micro-Touch® Sterile
Leverancier: ANSELL MEDICAL PRODUCTS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
553344 553343
112-1824EA 330 EUR
112-1824 112-1823
Onderzoekshandschoenen, latex, sterile, Micro-Touch® Sterile
Handschoenen
Micro-Touch® Sterile examination gloves are perfect for all medical applications where sterility is required and surgical gloves are not available (e.g. endoscopy, open wounds). Powder-free and with low levels of protein and residual chemicals, they minimise the risk of Type I allergies.
  • Powder-free, textured fingertips
  • Easy to don even with damp hands and provide excellent grip for handling instruments
  • Provide a secure barrier against viruses and micro-organisms
  • Interior polyacrylate coating makes the glove softer and more comfortable to wear
  • Gloves are thoroughly sterilised using gamma irradiation (minimum 25kGy)
Certificaten: EN 455; AQL 1,5
Verpakking: Packaged in pairs with a protective double wrapping.