4-Ethylphenol 97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
118820010 118820050 118822500
ACRO118820010EA 166 EUR
ACRO118820010 ACRO118820050 ACRO118822500
4-Ethylphenol 97%
4-Ethylphenol
Formule: C₈H₁₀O
Kookpunt: 218…219 °C (760 mmHg)
Smeltpunt: 41…46 °C
Dichtheid: 1,011 g/cm³ (20 °C)
Vlampunt: 100 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002393
CAS nummer: 123-07-9
EINECS: 204-598-6
UN: 2430
ADR: 8,III
Order Now
Specification Test Results
Appearance White to cream to brown Crystals or crystalline solid or fused mass
Infrared spectrum Conforms
Melting point 38 to 45 °C
GC ≥96.0 %