2-Propylphenol

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TL00268.1 TL00268.10 TL00268.50
MAYBTL00268.1EA 0 EUR
MAYBTL00268.1 MAYBTL00268.10 MAYBTL00268.50
2-Propylphenol
2-Propylphenol
Formule: C₉H₁₂O
Kookpunt: 224 °C (1013 hPa)
Smeltpunt: 7 °C
Dichtheid: 1,015 g/cm³ (0 °C)
MDL Nummer: MFCD00002251
CAS nummer: 644-35-9
EINECS: 211-415-3
UN: 3145
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR